1-Bromo-3,6-dimethoxy-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-3-fluoro-1,4-dimethoxybenzene

134302-1G 134302-5G 134302-25G
77464-630EA 65.92 USD
77464-630 77487-522 77498-314
1-Bromo-3,6-dimethoxy-2-fluorobenzene ≥95%
1-Bromo-3,6-dimethoxy-2-fluorobenzene

Precaución: For research use only

Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738437
CAS Number: 1783710-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR