5-Bromo-2-chloro-3-fluoroisonicotinic acid ≥95%

Supplier: AOBChem USA

Warning

133901-1G 133901-500MG 133901-5G 133901-25G
77464-626EA 419.21 USD
77464-626 77477-410 77487-518 77498-310
5-Bromo-2-chloro-3-fluoroisonicotinic acid ≥95%
5-Bromo-2-chloro-3-fluoroisonicotinic acid

Precaución: For research use only

Formula: C₆H₂BrClFNO₂
MW: 254.441 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29059810
CAS Number: 2091873-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR