(3-Fluoro-2,5-dimethylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

132424-1G 132424-500MG 132424-5G 132424-25G
77464-618EA 415.09 USD
77464-618 77477-400 77487-508 77498-300
(3-Fluoro-2,5-dimethylphenyl)(methyl)sulfane ≥95%
(3-Fluoro-2,5-dimethylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₁₁FS
MW: 170.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28794633
CAS Number: 1806333-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR