3-Ethoxy-5-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

132156-1G 132156-500MG 132156-5G 132156-25G
77464-614EA 262.65 USD
77464-614 77477-398 77487-506 77498-296
3-Ethoxy-5-fluorophenol ≥97%
3-Ethoxy-5-fluorophenol

Precaución: For research use only

Formula: C₈H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD29044442
CAS Number: 1443055-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR