6-Fluoro-4-(methylthio)pyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

131140-1G 131140-500MG 131140-5G 131140-25G
77464-610EA 82.4 USD
77464-610 77477-396 77487-502 77498-292
6-Fluoro-4-(methylthio)pyridin-3-ol ≥97%
6-Fluoro-4-(methylthio)pyridin-3-ol

Precaución: For research use only

Formula: C₆H₆FNOS
MW: 159.1813 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34828951
CAS Number: 2918968-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR