3-Bromo-2-fluoro-5-iodopyridine ≥95%

Supplier: AOBChem USA

Warning

13064-1G 13064-500MG 13064-5G 13064-25G
77464-606EA 245.14 USD
77464-606 77477-392 77487-496 77498-286
3-Bromo-2-fluoro-5-iodopyridine ≥95%
3-Bromo-2-fluoro-5-iodopyridine

Precaución: For research use only

Formula: C₅H₂BrFIN
MW: 301.883 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09038024
CAS Number: 697300-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR