2-Bromo-1-fluoro-3-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

130060-1G 130060-500MG 130060-5G 130060-25G
77464-602EA 351.23 USD
77464-602 77477-388 77487-490 77498-280
2-Bromo-1-fluoro-3-(trifluoromethoxy)benzene ≥95%
2-Bromo-1-fluoro-3-(trifluoromethoxy)benzene

Precaución: For research use only

Formula: C₇H₃BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD22551454
CAS Number: 1242249-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR