8-(2,5-Dimethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

130012-1G 130012-500MG 130012-5G 130012-25G
77464-598EA 397.58 USD
77464-598 77477-384 77487-486 77498-274
8-(2,5-Dimethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(2,5-Dimethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₆H₂₂O₃
MW: 262.344 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20862271
CAS Number: 1487364-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR