8-(4-(trifluoromethoxy)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

129990-1G 129990-500MG 129990-5G 129990-25G
77464-590EA 415.09 USD
77464-590 77477-376 77487-478 77498-266
8-(4-(trifluoromethoxy)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(4-(trifluoromethoxy)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₅H₁₇F₃O₄
MW: 318.288 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32879864
CAS Number: 151749-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR