8-(3-(dimethylamino)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129986-1G 129986-500MG 129986-5G 129986-25G
77464-586EA 415.09 USD
77464-586 77477-372 77487-474 77498-262
8-(3-(dimethylamino)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(3-(dimethylamino)phenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₆H₂₃NO₃
MW: 277.359 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32186441
CAS Number: 1252616-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR