8-(2-Chlorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129980-1G 129980-500MG 129980-5G 129980-25G
77464-582EA 397.58 USD
77464-582 77477-368 77487-470 77498-258
8-(2-Chlorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(2-Chlorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₄H₁₇ClO₃
Storage Temperature: Ambient
MDL Number: MFCD20862273
CAS Number: 1480156-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR