8-(3-Fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129974-1G 129974-500MG 129974-5G 129974-25G
77464-578EA 347.11 USD
77464-578 77477-364 77487-466 77498-254
8-(3-Fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(3-Fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₄H₁₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD21594348
CAS Number: 217476-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR