8-(3-Ethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129964-1G 129964-500MG 129964-5G
77464-574EA 497.49 USD
77464-574 77477-360 77487-462
8-(3-Ethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(3-Ethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₆H₂₂O₃
MW: 262.344 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23590195
CAS Number: 1518040-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR