8-(4-Ethoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129954-1G 129954-500MG 129954-5G 129954-25G
77464-570EA 397.58 USD
77464-570 77477-356 77487-458 77498-248
8-(4-Ethoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(4-Ethoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₆H₂₂O₄
Storage Temperature: Ambient
MDL Number: MFCD26743333
CAS Number: 163671-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR