8-(2-Ethoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129950-1G 129950-500MG 129950-5G 129950-25G
77464-566EA 415.09 USD
77464-566 77477-352 77487-454 77498-246
8-(2-Ethoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(2-Ethoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₆H₂₂O₄
Storage Temperature: Ambient
MDL Number: MFCD34828268
CAS Number: 1484061-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR