8-(3-Methoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129946-1G 129946-500MG 129946-5G 129946-25G
77464-562EA 397.58 USD
77464-562 77477-348 77487-450 77498-244
8-(3-Methoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(3-Methoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₅H₂₀O₄
Storage Temperature: Ambient
MDL Number: MFCD21594342
CAS Number: 365553-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR