8-(m-tolyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129940-1G 129940-500MG 129940-5G 129940-25G
77464-558EA 226.6 USD
77464-558 77477-344 77487-446 77498-240
8-(m-tolyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(m-tolyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₅H₂₀O₃
MW: 248.318 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21594335
CAS Number: 1252616-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR