4-Hydroxy-2,3,5-trifluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

12966-1G 12966-500MG 12966-5G 12966-25G
77464-554EA 148.32 USD
77464-554 77477-340 77487-442 77498-236
4-Hydroxy-2,3,5-trifluorobenzoic acid ≥95%
4-Hydroxy-2,3,5-trifluorobenzoic acid

Precaución: For research use only

Formula: C₇H₃F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD08458460
CAS Number: 156839-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR