3-(3-Fluorophenyl)-3-hydroxy-2-methylisoindolin-1-one ≥95%

Supplier: AOBChem USA

Warning

129620-1G 129620-500MG 129620-5G 129620-25G
77464-550EA 415.09 USD
77464-550 77477-336 77487-438 77498-232
3-(3-Fluorophenyl)-3-hydroxy-2-methylisoindolin-1-one ≥95%
3-(3-Fluorophenyl)-3-hydroxy-2-methylisoindolin-1-one

Precaución: For research use only

Formula: C₁₅H₁₂FNO₂
MW: 257.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34828168
CAS Number: 2969441-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR