2-(Methylamino)-5-pyrimidineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methylaminopyrimidine-5-boronic acid, pinacol ester, N-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

12958-1G 12958-500MG 12958-5G 12958-25G
77464-546EA 101.97 USD
77464-546 77477-332 77487-434 77498-228
2-(Methylamino)-5-pyrimidineboronic acid pinacol ester ≥97%
2-(Methylamino)-5-pyrimidineboronic acid pinacol ester

Precaución: For research use only

Formula: C₁₁H₁₈BN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD11878345
CAS Number: 904326-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR