1-(4-(trifluoromethoxy)phenyl)cyclooctanol ≥95%

Supplier: AOBChem USA

Warning

129564-1G 129564-500MG 129564-5G 129564-25G
77464-542EA 415.09 USD
77464-542 77477-328 77487-430 77498-224
1-(4-(trifluoromethoxy)phenyl)cyclooctanol ≥95%
1-(4-(trifluoromethoxy)phenyl)cyclooctanol

Precaución: For research use only

Formula: C₁₅H₁₉F₃O₂
MW: 288.305 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34828156
CAS Number: 2921873-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR