1-(3,5-bis(trifluoromethyl)phenyl)cyclooctanol ≥97%

Supplier: AOBChem USA

Warning

129558-1G 129558-500MG 129558-5G 129558-25G
77464-538EA 415.09 USD
77464-538 77477-324 77487-426 77498-220
1-(3,5-bis(trifluoromethyl)phenyl)cyclooctanol ≥97%
1-(3,5-bis(trifluoromethyl)phenyl)cyclooctanol

Precaución: For research use only

Formula: C₁₆H₁₈F₆O
MW: 340.304 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34828154
CAS Number: 2921866-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR