1-(2-Fluorophenyl)cyclooctanol ≥95%

Supplier: AOBChem USA

Warning

129537-1G 129537-500MG 129537-5G 129537-25G
77464-534EA 415.09 USD
77464-534 77477-320 77487-422 77498-216
1-(2-Fluorophenyl)cyclooctanol ≥95%
1-(2-Fluorophenyl)cyclooctanol

Precaución: For research use only

Formula: C₁₄H₁₉FO
MW: 222.298 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34828147
CAS Number: 2921904-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR