2,5-Dibromo-N-methylpyridin-3-amine ≥97%

Supplier: AOBChem USA

Warning

129525-1G 129525-500MG 129525-5G 129525-25G
77464-530EA 155.53 USD
77464-530 77477-316 77487-418 77498-212
2,5-Dibromo-N-methylpyridin-3-amine ≥97%
2,5-Dibromo-N-methylpyridin-3-amine

Precaución: For research use only

Formula: C₆H₆Br₂N₂
MW: 265.933 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723709
CAS Number: 2169192-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR