3-Chloro-5-fluoroisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-5-fluoro-4-pyridinecarboxylic acid

12923-1G 12923-500MG 12923-5G 12923-25G
77464-522EA 209.09 USD
77464-522 77477-308 77487-410 77498-206
3-Chloro-5-fluoroisonicotinic acid ≥97%
3-Chloro-5-fluoroisonicotinic acid

Precaución: For research use only

Formula: C₆H₃ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD13185823
CAS Number: 514798-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR