(2-Fluoro-3-(trifluoromethyl)phenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

128329-1G 128329-500MG 128329-5G 128329-25G
77464-518EA 415.09 USD
77464-518 77477-304 77487-406 77498-202
(2-Fluoro-3-(trifluoromethyl)phenyl)(furan-2-yl)methanol ≥95%
(2-Fluoro-3-(trifluoromethyl)phenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₂H₈F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD24930996
CAS Number: 1537726-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR