(2,5-Difluorophenyl)(furan-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

128323-1G 128323-500MG 128323-5G 128323-25G
77464-514EA 297.67 USD
77464-514 77477-300 77487-402 77498-198
(2,5-Difluorophenyl)(furan-2-yl)methanol ≥97%
(2,5-Difluorophenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₈F₂O₂
MW: 210.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787139
CAS Number: 1247238-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR