(3,5-bis(trifluoromethyl)phenyl)(furan-2-yl)methanol ≥96%

Supplier: AOBChem USA

Warning

128311-1G 128311-500MG 128311-5G 128311-25G
77464-510EA 415.09 USD
77464-510 77477-296 77487-398 77498-194
(3,5-bis(trifluoromethyl)phenyl)(furan-2-yl)methanol ≥96%
(3,5-bis(trifluoromethyl)phenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₃H₈F₆O₂
Storage Temperature: Ambient
MDL Number: MFCD34827572
CAS Number: 1538203-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR