1-(2,3-Difluorophenyl)cycloheptanol ≥95%

Supplier: AOBChem USA

Warning

128280-1G 128280-500MG 128280-5G 128280-25G
77464-506EA 415.09 USD
77464-506 77477-292 77487-394 77498-190
1-(2,3-Difluorophenyl)cycloheptanol ≥95%
1-(2,3-Difluorophenyl)cycloheptanol

Precaución: For research use only

Formula: C₁₃H₁₆F₂O
MW: 226.262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19626875
CAS Number: 1341463-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR