1-(4-Fluoro-2-methylphenyl)cycloheptanol ≥97%

Supplier: AOBChem USA

Warning

128274-1G 128274-500MG 128274-5G 128274-25G
77464-502EA 415.09 USD
77464-502 77477-288 77487-390 77498-186
1-(4-Fluoro-2-methylphenyl)cycloheptanol ≥97%
1-(4-Fluoro-2-methylphenyl)cycloheptanol

Precaución: For research use only

Formula: C₁₄H₁₉FO
MW: 222.298 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222260
CAS Number: 1343397-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR