1-(2,4-Difluorophenyl)cycloheptanol ≥97%

Supplier: AOBChem USA

Warning

128259-1G 128259-500MG 128259-5G 128259-25G
77464-498EA 415.09 USD
77464-498 77477-284 77487-386 77498-182
1-(2,4-Difluorophenyl)cycloheptanol ≥97%
1-(2,4-Difluorophenyl)cycloheptanol

Precaución: For research use only

Formula: C₁₃H₁₆F₂O
MW: 226.262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17223582
CAS Number: 1343751-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR