1-(3-(trifluoromethyl)phenyl)cycloheptanol ≥95%

Supplier: AOBChem USA

Warning

128253-1G 128253-500MG 128253-5G 128253-25G
77464-494EA 415.09 USD
77464-494 77477-280 77487-382 77498-178
1-(3-(trifluoromethyl)phenyl)cycloheptanol ≥95%
1-(3-(trifluoromethyl)phenyl)cycloheptanol

Precaución: For research use only

Formula: C₁₄H₁₇F₃O
MW: 258.279 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17389871
CAS Number: 39173-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR