1-(2-Fluorophenyl)cycloheptanol ≥96%

Supplier: AOBChem USA

Warning

128241-1G 128241-500MG 128241-5G 128241-25G
77464-490EA 415.09 USD
77464-490 77477-276 77487-378 77498-174
1-(2-Fluorophenyl)cycloheptanol ≥96%
1-(2-Fluorophenyl)cycloheptanol

Precaución: For research use only

Formula: C₁₃H₁₇FO
MW: 208.272 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16779260
CAS Number: 1250035-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR