1-(4-Fluorophenyl)cycloheptanol ≥97%

Supplier: AOBChem USA

Warning

128235-1G 128235-500MG 128235-5G 128235-25G
77464-486EA 297.67 USD
77464-486 77477-272 77487-374 77498-172
1-(4-Fluorophenyl)cycloheptanol ≥97%
1-(4-Fluorophenyl)cycloheptanol

Precaución: For research use only

Formula: C₁₃H₁₇FO
MW: 208.272 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021671
CAS Number: 77826-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR