3-Bromo-4-methyl-2-(methylthio)thiophene ≥95%

Supplier: AOBChem USA

Warning

127953-1G 127953-500MG 127953-5G 127953-25G
77464-482EA 415.09 USD
77464-482 77477-268 77487-368 77498-166
3-Bromo-4-methyl-2-(methylthio)thiophene ≥95%
3-Bromo-4-methyl-2-(methylthio)thiophene

Precaución: For research use only

Formula: C₆H₇BrS₂
Storage Temperature: Ambient
MDL Number: MFCD34827400
CAS Number: 2921838-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR