5-Cyclopentyl-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

127275-1G 127275-500MG 127275-5G
77464-474EA 889.92 USD
77464-474 77477-260 77487-360
5-Cyclopentyl-2-fluorobenzaldehyde ≥97%
5-Cyclopentyl-2-fluorobenzaldehyde

Precaución: For research use only

Formula: C₁₂H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD34826973
CAS Number: 2918952-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR