2-(2-Fluoro-4-iodophenyl)-2-methyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

127039-1G 127039-500MG 127039-5G 127039-25G
77464-470EA 415.09 USD
77464-470 77477-256 77487-354 77498-156
2-(2-Fluoro-4-iodophenyl)-2-methyl-1,3-dioxolane ≥97%
2-(2-Fluoro-4-iodophenyl)-2-methyl-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀FIO₂
MW: 308.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34826821
CAS Number: 2918964-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR