(1-Fluoronaphthalen-2-yl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

127029-1G 127029-500MG 127029-5G 127029-25G
77464-466EA 111.24 USD
77464-466 77477-254 77487-350 77498-152
(1-Fluoronaphthalen-2-yl)trimethylsilane ≥97%
(1-Fluoronaphthalen-2-yl)trimethylsilane

Precaución: For research use only

Formula: C₁₃H₁₅FSi
Storage Temperature: Ambient
MDL Number: MFCD34826813
CAS Number: 1873468-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR