(2,3-Difluoro-6-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

126820-1G 126820-500MG 126820-5G 126820-25G
77464-458EA 201.88 USD
77464-458 77477-246 77487-338 77498-142
(2,3-Difluoro-6-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%
(2,3-Difluoro-6-(methoxymethoxy)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀F₂O₂S
MW: 220.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34826717
CAS Number: 2918964-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR