4-Bromo-2-cyclopentyl-1-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

126502-1G 126502-500MG 126502-5G
77464-454EA 666.41 USD
77464-454 77477-242 77487-334
4-Bromo-2-cyclopentyl-1-fluorobenzene ≥95%
4-Bromo-2-cyclopentyl-1-fluorobenzene

Precaución: For research use only

Formula: C₁₁H₁₂BrF
MW: 243.115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26678746
CAS Number: 1541580-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR