1-(5-Bromo-2-fluorophenyl)propan-1-ol ≥97%

Supplier: AOBChem USA

Warning

126499-1G 126499-500MG 126499-5G 126499-25G
77464-450EA 341.96 USD
77464-450 77477-238 77487-330 77498-134
1-(5-Bromo-2-fluorophenyl)propan-1-ol ≥97%
1-(5-Bromo-2-fluorophenyl)propan-1-ol

Precaución: For research use only

Formula: C₉H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD13190783
CAS Number: 1197943-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR