6-Methyl-1H-indazol-3-amine ≥96%

Supplier: AOBChem USA

Warning

126095-1G 126095-500MG 126095-5G 126095-25G
77464-446EA 257.5 USD
77464-446 77477-234 77487-326 77498-128
6-Methyl-1H-indazol-3-amine ≥96%
6-Methyl-1H-indazol-3-amine

Precaución: For research use only

Formula: C₈H₉N₃
MW: 147.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07385617
CAS Number: 835616-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR