1-(5-Bromothiophen-2-yl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

125589-1G 125589-500MG 125589-5G
77464-442EA 576.8 USD
77464-442 77477-230 77487-322
1-(5-Bromothiophen-2-yl)-2-chloro-2,2-difluoroethanone ≥95%
1-(5-Bromothiophen-2-yl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₆H₂BrClF₂OS
MW: 275.498 g/mol
Storage Temperature: Ambient
CAS Number: 1824061-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR