1-Methyl-4-(thiophen-2-yl)piperidin-4-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
125574-1G
125574-500MG
125574-5G
125574-25G
77464-438EA
339.9
USD
77464-438
77477-226
77487-316
77498-120
1-Methyl-4-(thiophen-2-yl)piperidin-4-ol ≥97%
1-Methyl-4-(thiophen-2-yl)piperidin-4-ol
Precaución: For research use only
Formula:
C₁₀H₁₅NOS MW: 197.297 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01078463 CAS Number: 100131-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...