2-Fluoro-1-(thiophen-2-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

125570-1G 125570-500MG 125570-5G
77464-434EA 431.57 USD
77464-434 77477-222 77487-312
2-Fluoro-1-(thiophen-2-yl)ethanone ≥97%
2-Fluoro-1-(thiophen-2-yl)ethanone

Precaución: For research use only

Formula: C₆H₅FOS
MW: 144.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09054978
CAS Number: 371754-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR