4-(furan-2-yl)-1-methylpiperidin-4-ol ≥95%

Supplier: AOBChem USA

Warning

125555-1G 125555-500MG 125555-5G 125555-25G
77464-430EA 341.96 USD
77464-430 77477-218 77487-308 77498-116
4-(furan-2-yl)-1-methylpiperidin-4-ol ≥95%
4-(furan-2-yl)-1-methylpiperidin-4-ol

Precaución: For research use only

Formula: C₁₀H₁₅NO₂
MW: 181.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07774789
CAS Number: 75446-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR