5-Fluorobenzofuran-7-carbaldehyde ≥96%

Supplier: AOBChem USA

Warning

125542-1G 125542-500MG 125542-5G
77464-426EA 803.4 USD
77464-426 77477-212 77487-302
5-Fluorobenzofuran-7-carbaldehyde ≥96%
5-Fluorobenzofuran-7-carbaldehyde

Precaución: For research use only

Formula: C₉H₅FO₂
MW: 164.1332 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18822719
CAS Number: 555155-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR