8-(2-Chloro-6-fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

125484-1G 125484-500MG 125484-5G 125484-25G
77464-418EA 415.09 USD
77464-418 77477-206 77487-298 77498-110
8-(2-Chloro-6-fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(2-Chloro-6-fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₄H₁₆ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD34826188
CAS Number: 1854668-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR