8-(6-Bromo-2-fluoro-3-methoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥96%

Supplier: AOBChem USA

Warning

125471-1G 125471-500MG 125471-5G 125471-25G
77464-414EA 415.09 USD
77464-414 77477-202 77487-294 77498-106
8-(6-Bromo-2-fluoro-3-methoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥96%
8-(6-Bromo-2-fluoro-3-methoxyphenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₅H₁₈BrFO₄
MW: 361.2034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34826180
CAS Number: 2921892-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR