3-Bromo-2-(cyclohexyloxy)-5-methylpyridine ≥96%

Supplier: AOBChem USA

Warning

125414-1G 125414-500MG 125414-5G 125414-25G
77464-410EA 347.11 USD
77464-410 77477-198 77487-290 77498-104
3-Bromo-2-(cyclohexyloxy)-5-methylpyridine ≥96%
3-Bromo-2-(cyclohexyloxy)-5-methylpyridine

Precaución: For research use only

Formula: C₁₂H₁₆BrNO
MW: 270.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29114185
CAS Number: 1289024-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR